Anti-CD8a Mouse Monoclonal Antibody [clone: RPA-T8] (Brilliant Violet® 510)
Catalog # BLEG301048
Supplier: Biolegend
Specifications
- Antigen symbol:CD8a
- Clonality:Monoclonal
- Clone:RPA-T8
- Conjugation:Brilliant Violet® 510
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Baboon,Cynomolgus monkey,Sooty mangabey,Rhesus monkey,Pigtailed macaque,Chimpanzee,Human
- Antigen synonyms:T8|Leu2
- Storage buffer:Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and BSA (origin USA).
- Storage temperature:2…8 °C
- Tested applications:FC
- Purification:The antibody was purified by affinity chromatography and conjugated with Brilliant Violet 510™ under optimal conditions.
- Size:100 tests
- Pk:100 Tests