Anti-CD25 Mouse Monoclonal Antibody [clone: M-A251] (FITC (Fluorescein Isothiocyanate))
Supplier: Biolegend
Certificates
About this item
Specifications
- Catalog No:
- BLEG356105
- BLEG356106
- Antigen symbol:
- CD25
- CD25
- Antigen synonyms:
- IL-2Rα chain|Low affinity IL-2R|IL-2 receptor α chain
- IL-2Rα chain|Low affinity IL-2R|IL-2 receptor α chain
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Clone:
- M-A251
- M-A251
- Reactivity:
- Baboon, Cynomolgus monkey, Rhesus monkey, Human
- Baboon, Cynomolgus monkey, Rhesus monkey, Human
- Host:
- Mouse
- Mouse
- Isotype:
- IgG1 kappa
- IgG1 kappa
- Flow cytometry:
- Yes
- Yes
- Immunogen:
- Human PHA-induced lymphocyte cells
- Human PHA-induced lymphocyte cells
- Purification:
- The antibody was purified by affinity chromatography and conjugated with FITC under optimal conditions.
- The antibody was purified by affinity chromatography and conjugated with FITC under optimal conditions.
- Storage buffer:
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 0.2% (w/v) BSA (origin USA).
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 0.2% (w/v) BSA (origin USA).
- Storage temperature:
- 2…8 °C
- 2…8 °C
- Tested applications:
- FC
- FC
Specifications
Frequently Bought Together