Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
SFr. 0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Switzerland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Sulphuric acid
Sulphuric acid 100%, GPR RECTAPUR®
Sulphuric acid 100%, GPR RECTAPUR®
85741.243
:
VWR Chemicals
:
Sulphuric acid 100%, GPR RECTAPUR®
85741.243
:
VWR Chemicals
:
Pack type:
Glass bottle, PVC-coated
Pk:
2 Bottle
Seq:
5041
Pk Info:
2× 1 L
H₂SO₄ content (%):
98,2 - 100 %
:
:
98.08 g/mol
:
1830
:
8,II
:
GHS05
:
7664-93-9
:
H₂SO₄
:
MFCD00064589