Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
SFr. 0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Switzerland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Solvent S-316 for oil extraction
Solvent S-316 for oil extraction
Solvent S-316 for oil extraction
Solvent S-316 for oil extraction
Catalog # HORI5200100690
Supplier:
HORIBA
CAS Number:
Certificates
Expand image
Solvent S-316 for oil extraction
Catalog # HORI5200100690
Supplier:
HORIBA
CAS Number:
Certificates
Specifications
Pack type:
Glass bottle
Pk:
1,5 kg
Seq:
4046
Specifications
About this item
Product Details
Documentation
:
CAS Number:
9002-83-9
Product Details
Documentation