Order Entry
Switzerland
ContactUsLinkComponent
 
Human recombinant Tau-441 (2N4R)
  BLEG842501
 :  Biolegend
 :  
Human recombinant Tau-441 (2N4R)
  BLEG842501
 :  Biolegend
 :  

 

  • Pk:
    25 µG
  • Protein/peptide type:
    Recombinant
  • Species:
    Human
  • Storage conditions:
    –20…–70 °C
  • Protein synonyms:
    Isoform 4|Phf-Tau|Paired Helical Filament-Tau|Neurofibrillary Tangle Protein|G Protein Beta1/Gamma2 Subunit-Interacting Factor 1|Microtubule-Associated Protein Tau
  • Protein/peptide name:
    Tau-441 (2N4R)
  • Concentration:
    0,1 mg/ml
  • Formulation:
    pH 8,0, 50 mM TRIS, 300 mM NaCl, 1 mM TCEP, 1 mM EDTA, 10% Glycerol and protease inhibitors
  • Tested applications:
    WB, ELISA