Order Entry
Switzerland
ContactUsLinkComponent
Human recombinant HMGB1 (from 293E cells)
Human recombinant HMGB1 (from 293E cells)
Catalog # BLEG557804
Supplier:  Biolegend
CAS Number:  
Human recombinant HMGB1 (from 293E cells)
Catalog # BLEG557804
Supplier:  Biolegend
CAS Number:  

Specifications

  • Pk:
    25 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    293E cells
  • Species:
    Human
  • Storage conditions:
    –20…–70 °C
  • Carrier-free:
    Y
  • Protein synonyms:
    chromosomal protein non histone HMG1|amphoterin|HMG-1
  • Protein/peptide name:
    HMGB1
  • Concentration:
    0,2 mg/ml
  • Formulation:
    0,22 µm filtered protein solution is in PBS, pH 7,2
  • Tested applications:
    BA

Specifications

About this item