Order Entry
Switzerland
ContactUsLinkComponent
Mouse recombinant IL-25 (IL-17E) insect cells
Mouse recombinant IL-25 (IL-17E) insect cells
Supplier:  Biolegend
Certificates
Mouse recombinant IL-25 (IL-17E) insect cells
Supplier:  Biolegend

Specifications

  • Protein/peptide type:
    Recombinant
  • Source:
    Insect cells
  • Species:
    Mouse
  • Storage conditions:
    –20…–70 °C
  • Carrier-free:
    Y
  • Protein synonyms:
    IL17E|IL25
  • Protein/peptide name:
    IL-25 (IL-17E)
  • Concentration:
  • Formulation:
    0,22 µm filtered protein solution is in 20 mM NaHPO₄ pH 6,0, 0,5 M NaCl.
  • Tested applications:
    BA

Specifications

Frequently Bought Together