Order Entry
Switzerland
ContactUsLinkComponent
Human Recombinant TGF-beta1 (from CHO cells)
Human Recombinant TGF-beta1 (from CHO cells)
Catalog # BLEG781804
Supplier:  Biolegend
CAS Number:  
Human Recombinant TGF-beta1 (from CHO cells)
Catalog # BLEG781804
Supplier:  Biolegend
CAS Number:  

Specifications

  • Pk:
    25 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    CHO cells
  • Species:
    Human
  • Seq:
    02
  • Storage conditions:
    −20…−70 °C
  • Carrier-free:
    Y
  • Protein synonyms:
    TGF-Beta-1|transforming growth factor|Transforming Growth Factor Beta 1|TGFB|DPD1
  • Protein/peptide name:
    TGF-beta1
  • Concentration:
    0,2 mg/ml
  • Formulation:
    0,22 µm filtered protein solution is in <30% Acetonitrile, 0,1% TFA (trifluoroacetic acid).
  • Tested applications:
    BA

Specifications

About this item