Human Recombinant VEGF
Catalog # BLEG560309
Supplier: Biolegend
CAS Number:
Specifications
- Pk:4
- Protein/peptide type:Recombinant
- Species:Human
- Storage conditions:2…8 °C
- Protein synonyms:vascular endothelial growth factor|MVCD1|VEGFA|VEGF-A|VPF
- Protein/peptide name:VEGF
- Formulation:Lyophilized in sterile-filtered PBS, pH 7,2, containing 1% BSA, 0,09% sodium azide, and protease inhibitors.
- Tested applications:ELISA