Order Entry
Switzerland
ContactUsLinkComponent
Mouse Recombinant IFN-alpha (from E. coli)
Mouse Recombinant IFN-alpha (from E. coli)
Supplier:  Biolegend
Certificates
Mouse Recombinant IFN-alpha (from E. coli)
Supplier:  Biolegend

Specifications

  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Mouse
  • Storage conditions:
    –20…–70 °C
  • Carrier-free:
    Y
  • Protein synonyms:
    IFN-a|IFNa|MuIFN-alpha-A|Ifnaa|Mus musculus interferon alpha 15 (Ifna15)|IFN-alpha
  • Protein/peptide name:
    IFN-alpha
  • Concentration:
    0,2 mg/ml
  • Formulation:
    0,22 µm filtered protein solution is in PBS, pH 7,2.
  • Tested applications:
    BA

Specifications

Frequently Bought Together