Order Entry
Switzerland
ContactUsLinkComponent
Human recombinant visfatin
Human recombinant visfatin
Supplier:  Biolegend
Certificates
Human recombinant visfatin
Supplier:  Biolegend

Specifications

  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Storage conditions:
    –20...–70 °C
  • Carrier-free:
    Y
  • Protein synonyms:
    Nicotinamide phosphoribosyltransferase|PBEF (Pre-B cell colony-enhancing factor)
  • Protein/peptide name:
    Visfati
  • Formulation:
    Lyophilized
  • Tested applications:
    BA

Specifications

Frequently Bought Together