ContactUsLinkComponent
QUANTOFIX Copper Sensitive 5
QUANTOFIX Copper Sensitive 5
  MANA91358
 :  
undefined
QUANTOFIX Copper Sensitive 5
  MANA91358
 :  
  • Pk:
    25
  • Description:
    QUANTOFIX Copper Sensitive 5, 25 tests
Semi-quantitative test strips for reliable determination of copper concentrations as low as 0.1 mg/L (ppm). A hole in the plastic carrier allows the water to migrate through the test pad increasing the sensitivity significantly. Ideal for rapid and easy analysis of copper directly on-site. No maintenance or calibration is required.
  • Sensitive rapid test