Order Entry
Switzerland
ContactUsLinkComponent
VWR® Trolleys for Euro Containers
Catalog # 216-2439
Supplier:  VWR Collection
undefined
VWR® Trolleys for Euro Containers
Catalog # 216-2439
Supplier:  VWR Collection

Specifications

  • Colour:
    Grey
  • Description:
    Brakeless trolley for 800×600 container, ABS material, 250 kg load capacity, Ø125 mm wheel
  • Seq:
    02
  • W×D×H:
    820×620×200 mm
  • Pk:
    1

Specifications

About this item

Transport trolleys made from ABS.