Fine dosage syringes, Injekt® Luer Duo
Supplier: B. Braun
Certificates
About this item
2-piece hypodermic syringes with Luer connector for single use with detached hypodermic Sterican® needle.
- Green plunger for precise dosage
- Highly transparent barrel with black graduation for easy reading
- Safe plunger backstop to prevent unintentional plunger withdrawal
- Graduation extended beyond
- nominal volume
- Not manufactured with silicone oil,
- PVC, Latex and DEHP
Barrel made of PP, plunger made of PE.
Packaging: Sterile single packaging in box of 100 units.
Frequently Bought Together