Celite® Hyflo Super-cel®, filter aid, GPR RECTAPUR®
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:68855-54-9
:
MDL Number:MFCD00132803
Melting Pt:>450 °C
Density:2.36 g/cm³ (20 °C)
REACH:01-2119488518-22-XXXX
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Identification :Passes test
- pH (25 °C; 10 %) 8.0 - 11.0:8,0 - 11,0
- Wet density ≤ 353 g/l:≤ 353 g/l
- Loss on ignition (600 °C) :≤ 0.5 %
- Particle size (< 0.1 mm) :≥ 85 %
Specification Test Results
Related Information
Certificates
Search by Lot Numberspec_VWRC22559.290 spec_VWRC22559.360
Product specification
VWR® for Drying and Filtration
Product Literature
Frequently Bought Together