Order Entry
Switzerland
ContactUsLinkComponent
Sepharose Chromatography Resins, Strep-Tactin® XT
Sepharose Chromatography Resins, Strep-Tactin® XT
Supplier:  Cytiva
Certificates
undefined
Sepharose Chromatography Resins, Strep-Tactin® XT
Supplier:  Cytiva

Specifications

  • pH range:
    6 - 10
  • Matrix:
    Rigid, cross-linked agarose, spherical
  • Ligand:
    Strep-Tactin XT
  • Storage temperature:
    2...8 °C in 20% ethanol
  • Binding capacity:
    ~10 mg protein with Strep-tag II or Twin-Strep-tag/ml of resin
  • Ligand density:
    ~5 mg/ml resin
  • Flow rate:
    300 cm/h

Specifications

Frequently Bought Together