Paq5000 DNA polymerase
Anbieter: AGILENT
Zertifikate
Über diesen Artikel
Paq5000 is capable of quickly and robustly amplifying up to 6 kB genomic DNA targets.
- Equal to better yield with fast cycling conditions
- Robust, cost-effective
- Pre-optimised reaction buffers included
- Low cost hot start formulation
With a shortened extension rate of 30 sec/kB and robust yields, Paq5000 is a Taq alternative that can be substituted easily into many Taq-based procedures.
Paq5000 includes the ArchaeMaxx factor, improving the yield of products by overcoming dUTP poisoning, which is caused by dUTP accumulation during PCR through dCTP deamination. Once incorporated, dU-containing DNA inhibits the enzyme and the ArchaeMaxx factor functions as a dUTPase, converting poisonous dUTP to harmless dUMP and inorganic pyrophosphate.
Caution: For research use only. Not for use in diagnostic procedures.
Frequently Bought Together