Orders LinkContactUsLinkComponent
pBK-CMV phagemid vector
pBK-CMV phagemid vector
pBK-CMV phagemid vector, 20 µG
Catalog # AGLS212209
Supplier:   AGILENT
pBK-CMV phagemid vector
Catalog # AGLS212209
Supplier:  AGILENT
CAS Number:  
pBK-CMV phagemid vector
pBK-CMV phagemid vector, 20 µG
CAS Number:  
Price
Inquire For Price
Log in for availability
1 * 20 µG
Product Details & Documents
pBK-CMV vector used to express proteins in both E. coli and mammalian model systems.
  • Constitutive expression in eukaryotic cells
  • Inducible protein expression in prokaryotes
  • Rescued from ZAP Express lambda vector
In mammalian cells, the expression is driven by the CMV promoter and is therefore constitutively expressed. In E. coli, the lac promoter drives high-level expression in the presence of the inducer IPTG, and allows minimal expression in the absence of inducer.
Allows both prokaryotic and eukaryotic expression.
Delivery information: Kit contains 20 μg pBK-CMV phagemid vector, 500 μl XL1-Blue MRF, and 1 ml R408 Interference-Resistance helper phage.
Caution: For research use only. Not for use in diagnostic procedures.
Specifications
Specifications
  • Pk:
    20 µG
  • Description:
    pBK-CMV phagemid vector