Orders LinkContactUsLinkComponent
Cryoboxes, Corning®
Cryoboxes, Corning®
Supplier:   Corning
Cryoboxes, Corning®
Supplier:  Corning
Certificates
Cryoboxes, Corning®
Cryoboxes, Corning®
3 Options Available Below
Product Details & Documents
PC, autoclavable.
  • Reusable
  • Ridges on the sides of the box make them easier to hold while wearing gloves
  • Transparent lid with numeric grid
Designed for use with most cryogenic vials.
These boxes feature a large marking spot and a clear polycarbonate lid that can be labelled to show vial contents. They are stackable and are available in two formats: 81-place boxes which will hold internally and externally threaded vials; and 100 count boxes, which will hold internally threaded vials.
These cryogenic storage boxes are suitable for storage down to –196 °C (–320,8 °F).
Caution: *Accepts internally threaded cryogenic vials only
Product Family Options
Product Information
  • Compartments
    For
    Availability
    Price
  • 9×9
    Cryovials 1 - 2 ml
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - SFr. 278.00
    SFr. 278.00
    1 * 10 ST
       SFr. 278.00
  • 9×9
    Cryovials 4 - 5 ml
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - SFr. 409.00
    SFr. 409.00
    1 * 10 ST
       SFr. 409.00
  • 10×10
    Cryovials 1 - 2 * ml
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - SFr. 321.00
    SFr. 321.00
    1 * 10 ST
       SFr. 321.00