ContactUsLinkComponent
Capmats for J.T.Baker® Plate Round Well
Capmats for J.T.Baker® Plate Round Well
N° de catalogue 43001-0017
Fournisseur:  Avantor
   Low Manufacturing Impact
undefined
Capmats for J.T.Baker® Plate Round Well
N° de catalogue 43001-0017
Fournisseur:  Avantor
   Low Manufacturing Impact
Spécifications
  • Optical property:
    Transparent
  • Packed:
    1 bag of 20
  • Sterile:
    -
  • Description:
    Capmat for J.T.Baker Plate Round Well
  • Material:
    EVA
  • Environmentally Preferable:
    Low Manufacturing Impact
  • Environmental certification:
    ISO 50001 (Certificate Registration No. 10000406726-MSC-RvA-DEU)
  • Pk:
    20
Spécifications
À propos de cet article
Suitable to use with J.T.Baker® Plate Round Well 43001-0010, 43001-0016 or 43001-0116.