ContactUsLinkComponent
Multiple affinity removal spin cartridge HSA/IgG
Multiple affinity removal spin cartridge HSA/IgG
Catalog # AGLS5188-8825
Supplier:  AGILENT
Multiple affinity removal spin cartridge HSA/IgG
Catalog # AGLS5188-8825
Supplier:  AGILENT
Specifications
  • Capacity:
    0,45 ml
  • Description:
    Multiple affinity removal system spin cartridge, human albumin/IgG
  • Pk:
    1
Specifications
About this item
The multiple affinity removal spin cartridge – Human Albumin/IgG is designed to remove two interfering high-abundant proteins from human samples (such as plasma, serum, urine or cerebrospinal fluid).
  • Compatible with standard benchtop microcentrifuges and luer-lok syringes
  • Serum load volume is 50 µl per run
  • Optimise sample recovery by utilising our rapid, single injection protocols and optimised buffers
Removal of these abundant proteins improves the subsequent LC/MS and electrophoretic analysis of the sample by effectively expanding the dynamic range of the analysis.
Caution: For research use only. Not for use in diagnostic procedures.