ContactUsLinkComponent
SIPS 10/20 pump tubing
SIPS 10/20 pump tubing
Catalog # VARI9910076100
Supplier:  VARIAN
SIPS 10/20 pump tubing
Catalog # VARI9910076100
Supplier:  VARIAN
Specifications
  • Description:
    Tubing kit for SIPS 10/20, to connect constant pressure head bottle to tee piece
  • Pk:
    1
Specifications
About this item
SIPS 10/20 pump tubing for atomic absorption spectroscopy.
  • Selected to provide extended lifetime and accurate dilution when using the SIPS
The Sample Introduction Pump System (SIPS) is an innovative sample introduction and dilution system for flame AA that eliminates the task of manually preparing multiple calibration standards, providing fast, accurate on-line dilution of over range samples.