Order Entry
Switzerland
ContactUsLinkComponent
AdEasy virus purification kit
AdEasy virus purification kit
Catalog # AGLS240243
Supplier:  AGILENT
CAS Number:  
undefined
AdEasy virus purification kit
Catalog # AGLS240243
Supplier:  AGILENT
CAS Number:  

Specifications

  • Pk:
    1 kt
  • Refine target molecule:
    Viral DNA
  • Target molecule:
    DNA
  • Description:
    AdEasy virus purification kit

Specifications

About this item

The AdEasy Virus Purification Kit allows the purification and concentration of Adenovirus (from Ad5 strains) with Sartobind® syringe filters containing an ion exchange membrane adsorber that selectively binds adenoviral particles. Once bound, viral particles can be further purified by washing away nonspecifically-bound proteins before elution. Concentrated and purified viral particles can be obtained in 2 to 3 hours, in contrast to traditional CsCl gradient centrifugation which typically takes 12 to 48 hours.

  • Rapid alternative to cesium chloride (CsCl) density gradient methods
  • No cesium chloride or ultracentrifugation necessary
  • Increased infection rates and cell viability

AdEasy virus purification kits rapid membrane chromatography is based method for the purification and concentration of type 5 adenoviral stocks.

Caution: For Research Use Only. Not for Use in Diagnostic Procedures.