Orders LinkContactUsLinkComponent
Solid phase extraction cartridges, Bond Elut™ CN-U
Solid phase extraction cartridges, Bond Elut™ CN-U
Supplier:   VARIAN
Solid phase extraction cartridges, Bond Elut™ CN-U
Supplier:  VARIAN
Certificates
Solid phase extraction cartridges, Bond Elut™ CN-U
4 Options Available Below
Product Details & Documents
Bond Elut CN-U is ideally suited for the extraction of polar compounds from a nonpolar matrix, such as hexane or oils. It is suitable for very polar analytes that may be irreversibly retained on SI or Diol (2OH) SPE phases.
  • Ideal for extracting aqueous analytes
  • Retention in aqueous and organic matrices
  • Useful for many applications
A medium polarity sorbent with many uses, it is ideal for applications in which extremely nonpolar compounds would be irreversibly retained on high carbon load sorbents, such as C8 and C18. This endcapped version of the cyano sorbent is best utilized when extracting analytes from an aqueous matrix.
Product Family Options
Bond Elut™ Jr cartridge
Product Information
  • Phase
    Particle Size
    Capacity
    Bed weight
    Availability
    Price
  • VARI12166053B
    Bond Elut™ Jr cartridge
    Bond Elut™ Jr cartridge
    Cyano
    40 µm
    1000 mg
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST
LRC cartridges
Product Information
  • Phase
    Particle Size
    Capacity
    Bed weight
    Availability
    Price
  • VARI12113034
    LRC cartridge
    LRC cartridge
    Cyano
    40 µm
    10 ml
    500 mg
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST
Straight barrel cartridges
Product Information
  • Phase
    Particle Size
    Capacity
    Bed weight
    Availability
    Price
  • VARI12102066
    Straight barrel cartridge
    Straight barrel cartridge
    Cyano
    40 µm
    1 ml
    50 mg
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST
  • 551-0304
    Straight barrel cartridge
    Straight barrel cartridge
    Cyano
    40 µm
    1 ml
    100 mg
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST