Syringe filters, SupaTop
: GILSON
SupaTop syringe filters offer reliable and cost-effective solutions for particles removal and clarification needs.
- Body made of pigment-free PP
- Certified for low levels of UV-absorbing extractables
A large variety of membranes are available for all applications: MCE, Nylon, PTFE, PES or PVDF.
Frequently Bought Together