ContactUsLinkComponent
HRP-conjugate stabiliser, SeramunStab® Stplus
HRP-conjugate stabiliser, SeramunStab® Stplus
Catalog # APOSBIL3111-100ML
Supplier:  Apollo Scientific
CAS Number:  
HRP-conjugate stabiliser, SeramunStab® Stplus
Catalog # APOSBIL3111-100ML
Supplier:  Apollo Scientific
CAS Number:  
Specifications
  • Pk:
    100 mL
  • Description:
    SeramunStab® Stplus HRP-conjugate stabiliser
  • Seq:
    01
Specifications
About this item
Liquid product colourless to slightly yellow or greenish, odourless, pH 7,3 to 7,5. Stabiliser for conjugates with monomeric HRP.