Order Entry
Switzerland
Orders LinkContactUsLinkComponent
DNA ligation kit

DNA ligation kit

DNA ligation kit, 150, 1 KIT
Catalog # AGLS203003
Supplier:   AGILENT
DNA ligation kit
Catalog # AGLS203003
Supplier:  AGILENT
CAS Number:  
DNA ligation kit

DNA ligation kit, 150, 1 KIT

CAS Number:  
Price
Inquire For Price
Log in for availability
1 * 1 KIT
Product Details & Documents

The DNA Ligation Kit includes T4 DNA ligase for highly efficient ligations.

  • For both sticky-end and blunt-end ligations
  • Reagents guaranteed to be DNase free while providing optimal ligation efficiency
  • Includes a plasmid control and a lambda control

The kit is optimised for use with lambda arms and plasmid vectors.

Delivery information: Kit contains T4 DNA ligase (300 U) 10X ligase buffer plasmid control Lambda control 10 mM rATP.

Caution: For research use only. Not for use in diagnostic procedures.

Specifications

Specifications

  • Pk:
    1 KIT
  • Description:
    DNA ligation kit
  • No. of reactions:
    150

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.