Orders LinkContactUsLinkComponent
Big blue
Big blue
Transpack packaging extract, 400 reactions, 1
Catalog # AGLS200220
Supplier:   AGILENT
Big blue
Catalog # AGLS200220
Supplier:  AGILENT
CAS Number:  
Big blue
Transpack packaging extract, 400 reactions, 1
CAS Number:  
Price
SFr. 29’776.00
Log in for availability
1 * 1 ST
Product Details & Documents
These are used in all Big Blue rodents and cell lines.
Transpack packaging extract is optimised for use with Big Blue Rodents and Cell lines as well as the lambda CII select Assay.
The Big Blue transgenic rodent mutagenesis assay is an in vivo system for assessing the genetic toxicity of test compounds.
The Lambda Select-cII mutation detection system for Big Blue rodents is a positive mutant-screening method for mutagenesis studies using Big Blue transgenic rodents.
Big Blue paque colour controls with four separate phage suspensions of colour mutants mixed with non-mutant phage.
Specifications
Specifications
  • Pk:
    1
  • Description:
    Transpack packaging extract
  • Tests per kit:
    400 reactions