Orders LinkContactUsLinkComponent
Deoxynucleotide mix
Deoxynucleotide mix
Deoxynucleotide mix, PCR-grade, 0,4 ml, 1
Catalog # AGLS200415
Supplier:   AGILENT
Deoxynucleotide mix
Catalog # AGLS200415
Supplier:  AGILENT
CAS Number:  
Deoxynucleotide mix
Deoxynucleotide mix, PCR-grade, 0,4 ml, 1
CAS Number:  
Price
SFr. 353.00
Log in for availability
1 * 1 ST
Product Details & Documents
The PCR grade deoxynucleotide mix includes 25 mM of each standard dNTP (GATC) for use with any PCR application, including real-time quantitative PCR.
  • Ideal for optimal PCR performance
  • Produces high-quality reaction products
  • Withstands multiple freeze-thaw without compromising efficiency
Delivery information: Includes 400 µl of 100 mM dNTP mix (25 mM of each dNTP), adequate for 1000 standard 50 µl primer-extension reactions or 2000 reactions of 25 µl.
Specifications
Specifications
  • Pk:
    1
  • Description:
    Deoxynucleotide mix, PCR-grade, 0,4 ml