Order Entry
Switzerland
ContactUsLinkComponent
Genomic DNA isolation
Genomic DNA isolation
Supplier:  AGILENT
Certificates
Genomic DNA isolation
Supplier:  AGILENT

Specifications

  • Method / Format:
    Precipitation
  • Prep size:
    Scalable
  • Refine target molecule:
    Total DNA
  • Sample type:
    Cultured cells
    Human/animal cells and tissues
  • Target molecule:
    DNA

Specifications

Frequently Bought Together