QuikChange HT protein engineering system
About this item
Protein directed evolution in just one step.
- Discover fast, learn more
- T300Quickly identify functional sites-codon saturation mutagenesis across the entire protein
- Efficient design reduces screening costs
- Only the mutations you want with all mutants retaining structural integrity
- Time and cost savings
- From DNA to colonies in less than 24 hours with QuikChange Lightning speed
The QuikChange HT protein engineering system provides rapid resolution of structural and functional questions by creating libraries of rationally designed mutants for applications such as single amino acid scanning, site saturations scanning, or targeted combinatorial mutagenesis.
Precisely target and mutate some or each position in 1 to 20×50 amino acid regions in less than 24 hours. Design large mutagenic libraries from as few as 100 and up to 120000 user-defined sequences with free access to mutagenesis workspace software in eArray. Discover fast, learn more.
QuikChange HT Protein Engineering System contains up to 120000 mutagenic oligonucleotides, oligo amplification primers and reagents, QuikChange enzymes and reagents, and SoloPack Gold SuperCompetent cells for EZClone.
Caution: For research use only. Not for use in diagnostic procedures.
8 Options Available Below
Product Details & Documents
Protein directed evolution in just one step.
- Discover fast, learn more
- T300Quickly identify functional sites-codon saturation mutagenesis across the entire protein
- Efficient design reduces screening costs
- Only the mutations you want with all mutants retaining structural integrity
- Time and cost savings
- From DNA to colonies in less than 24 hours with QuikChange Lightning speed
The QuikChange HT protein engineering system provides rapid resolution of structural and functional questions by creating libraries of rationally designed mutants for applications such as single amino acid scanning, site saturations scanning, or targeted combinatorial mutagenesis.
Precisely target and mutate some or each position in 1 to 20×50 amino acid regions in less than 24 hours. Design large mutagenic libraries from as few as 100 and up to 120000 user-defined sequences with free access to mutagenesis workspace software in eArray. Discover fast, learn more.
QuikChange HT Protein Engineering System contains up to 120000 mutagenic oligonucleotides, oligo amplification primers and reagents, QuikChange enzymes and reagents, and SoloPack Gold SuperCompetent cells for EZClone.
Caution: For research use only. Not for use in diagnostic procedures.
Product Family Options
Product Information
- PkAvailabilityPrice
- AGLSG5900AQuikChange HT Protein engineering system 150nt, 10 SitesQuikChange HT Protein engineering system 150nt, 10 Sites1 ktLog in for availability1 * 1 ktSFr. 15’684.00
- AGLSG5900BQuikChange HT Protein engineering system 150nt, 20 SitesQuikChange HT Protein engineering system 150nt, 20 Sites1 ktLog in for availability1 * 1 ktSFr. 24’130.00
- AGLSG5901AQuikChange HT Protein engineering system 200nt, 10 SitesQuikChange HT Protein engineering system 200nt, 10 Sites1 ktLog in for availability1 * 1 ktSFr. 19’127.00
- AGLSG5901BQuikChange HT Protein engineering system 200nt, 20 SitesQuikChange HT Protein engineering system 200nt, 20 Sites1 ktLog in for availability1 * 1 ktSFr. 29’427.00
- AGLSG5902AQuikChange HT Protein engineering system 150nt-Academic, 10 SitesQuikChange HT Protein engineering system 150nt-Academic, 10 Sites1 ktLog in for availability1 * 1 ktSFr. 9’294.00
- AGLSG5902BQuikChange HT Protein engineering system 150nt-Academic, 20 SitesQuikChange HT Protein engineering system 150nt-Academic, 20 Sites1 ktLog in for availability1 * 1 ktSFr. 14’299.00
- AGLSG5903AQuikChange HT Protein engineering system 200nt-Academic, 10 SitesQuikChange HT Protein engineering system 200nt-Academic, 10 Sites1 ktLog in for availability1 * 1 ktSFr. 11’335.00
- AGLSG5903BQuikChange HT Protein engineering system 200nt-Academic, 20 SitesQuikChange HT Protein engineering system 200nt-Academic, 20 Sites1 ktLog in for availability1 * 1 ktSFr. 17’438.00