Order Entry
Switzerland
ContactUsLinkComponent
Recombinant staphylokinase
Recombinant staphylokinase
Catalog # APOSBITP1348-1MG
Supplier:  Apollo Scientific
CAS Number:  
Recombinant staphylokinase
Catalog # APOSBITP1348-1MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pk:
    1 mg
  • Enzyme type:
    Recombinant
  • Source:
  • Species:
  • Environmentally Preferable:
  • Storage conditions:
    Ambient
  • MDL No.:
    MFCD09702076
  • Enzyme name:
    Staphylokinase
  • Enzyme synonyms:
    Staphylokinase

Specifications

About this item