Order Entry
Switzerland
ContactUsLinkComponent
Anti-CD62E Mouse Antibody
Anti-CD62E Mouse Antibody
Catalog # APOSBITP2133-1MG
Supplier:  Apollo Scientific
Anti-CD62E Mouse Antibody
Catalog # APOSBITP2133-1MG
Supplier:  Apollo Scientific

Specifications

  • Antigen name:
    E-Selectin
  • Antigen symbol:
    CD62E
  • Host:
    Mouse
  • Reactivity:
    Human
  • Pk:
    1 mg

Specifications

About this item