Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
SFr. 0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Switzerland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Antibodies
Anti-CD4 Mouse Antibody
Anti-CD4 Mouse Antibody
Katalognr. APOSBITP2004-1MG
Anbieter:
Apollo Scientific
Bild vergrößern
Anti-CD4 Mouse Antibody
Katalognr. APOSBITP2004-1MG
Anbieter:
Apollo Scientific
Technische Daten
Antigen symbol:
CD4
Host:
Mouse
Reactivity:
Human
Pk:
1 mg
Technische Daten
Über diesen Artikel
Produktdetails
Produktdetails