Order Entry
Switzerland
ContactUsLinkComponent
Seabream recombinant GH
Seabream recombinant GH
Catalog # APOSBITP2083-1MG
Supplier:  Apollo Scientific
CAS Number:  
Seabream recombinant GH
Catalog # APOSBITP2083-1MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pk:
    1 mg
  • Protein/peptide type:
    Recombinant
  • Species:
    Sea bream
  • Protein synonyms:
    Gilthead Seabream Growth Hormone
  • Protein/peptide name:
    GH

Specifications

About this item