Order Entry
Switzerland
ContactUsLinkComponent
 
Recombinant HIV-1 gag p17, p24
Catalog # APOSBITP1888-1MG
Supplier:  Apollo Scientific
CAS Number:  
Recombinant HIV-1 gag p17, p24
Catalog # APOSBITP1888-1MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pk:
    1 mg
  • Protein/peptide type:
    Recombinant
  • Protein synonyms:
    HIV-1 p17/24
  • Protein/peptide name:
    HIV-1 gag p17, p24

Specifications

About this item