Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
SFr. 0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Switzerland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Proteins and Peptides
Recombinant HIV-1 Envelope( 233)
Recombinant HIV-1 Envelope( 233)
Recombinant HIV-1 Envelope( 233)
APOSBITP1864-1MG
:
Apollo Scientific
:
Recombinant HIV-1 Envelope( 233)
APOSBITP1864-1MG
:
Apollo Scientific
:
Pk:
1 mg
Protein/peptide type:
Recombinant
Protein synonyms:
HIV-1Envelope
Protein/peptide name:
HIV-1 Envelope( 233)