Order Entry
Switzerland
ContactUsLinkComponent
Recombinant Varicella Zoster Virus gE
Recombinant Varicella Zoster Virus gE
Catalog # APOSBITP1846-1MG
Supplier:  Apollo Scientific
CAS Number:  
Recombinant Varicella Zoster Virus gE
Catalog # APOSBITP1846-1MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pk:
    1 mg
  • Protein/peptide type:
    Recombinant
  • Protein synonyms:
    VZV gE
  • Protein/peptide name:
    Varicella Zoster Virus gE

Specifications

About this item