Recombinant Hepatitis E Virus ORF2 (403-461)
Catalog # APOSBITP1829-1MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pk:1 mg
- Protein/peptide type:Recombinant
- Protein synonyms:HEV ORF2
- Protein/peptide name:Hepatitis E Virus ORF2 (403-461)