Recombinant Hepatitis C Virus NS5 Genotype-4
Catalog # APOSBITP1804-1MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pk:1 mg
- Protein/peptide type:Recombinant
- Protein synonyms:HCV NS5 Genotype-4
- Protein/peptide name:Hepatitis C Virus NS5 Genotype-4