Order Entry
Switzerland
ContactUsLinkComponent
Recombinant Hepatitis C Virus NS5 Genotype-1
Recombinant Hepatitis C Virus NS5 Genotype-1
Catalog # APOSBITP1795-1MG
Supplier:  Apollo Scientific
CAS Number:  
Recombinant Hepatitis C Virus NS5 Genotype-1
Catalog # APOSBITP1795-1MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pk:
    1 mg
  • Protein/peptide type:
    Recombinant
  • Protein synonyms:
    HCV NS5 Genotype-1
  • Protein/peptide name:
    Hepatitis C Virus NS5 Genotype-1

Specifications

About this item