Recombinant Hepatitis C Virus NS4 a+b, Fluorescein
Catalog # APOSBITP1792-1MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pk:1 mg
- Conjugation:Fluorescein
- Protein/peptide type:Recombinant
- Protein synonyms:HCV NS4 a+b
- Protein/peptide name:Hepatitis C Virus NS4 a+b