SFr. 0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Switzerland
ContactUsLinkComponent
Proteins and Peptides
Recombinant Der P1 Protein
Recombinant Der P1 Protein
APOSBITP1723-1MG
:
Apollo Scientific
:
Recombinant Der P1 Protein
APOSBITP1723-1MG
:
Apollo Scientific
:
Pk:
1 mg
Protein/peptide type:
Recombinant
Protein synonyms:
Der-1
Protein/peptide name:
Der P1 Protein