Human Recombinant TNF-a M
Catalog # APOSBITP1286-1MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pk:1 mg
- Protein/peptide type:Recombinant
- Species:Human
- Protein synonyms:Tumor Necrosis Factor-alpha mutant/variant
- Protein/peptide name:TNF-a M