SFr. 0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Switzerland
ContactUsLinkComponent
Proteins and Peptides
Human Recombinant IGF-I
Human Recombinant IGF-I
APOSBITP1106-1MG
:
Apollo Scientific
:
Human Recombinant IGF-I
APOSBITP1106-1MG
:
Apollo Scientific
:
Pk:
1 mg
Protein/peptide type:
Recombinant
Species:
Human
Protein synonyms:
Insulin Like Growth Factor-I
Protein/peptide name:
IGF-I