Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
SFr. 0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Switzerland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Proteins and Peptides
Human recombinant MIG
Human recombinant MIG
Catalog # APOSBITC2031-1MG
Anbieter:
Apollo Scientific
CAS-Nummer:
Expand image
Human recombinant MIG
Catalog # APOSBITC2031-1MG
Anbieter:
Apollo Scientific
CAS-Nummer:
Technische Daten
Pk:
1 mg
Protein/peptide type:
Recombinant
Species:
Human
Protein synonyms:
CXCL9
Protein/peptide name:
MIG
Technische Daten
Über diesen Artikel
Product Details
Product Details