Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
SFr. 0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Switzerland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Proteins and Peptides
Rat recombinant rRMCP-1
Rat recombinant rRMCP-1
APOSBITC2026-1MG
:
Apollo Scientific
:
Rat recombinant rRMCP-1
APOSBITC2026-1MG
:
Apollo Scientific
:
Pk:
1 mg
Protein/peptide type:
Recombinant
Species:
Rat
Protein synonyms:
Monocyte Chemotactic Protein-1, CCL2
Protein/peptide name:
rRMCP-1