Order Entry
Switzerland
ContactUsLinkComponent
Mouse recombinant GRO-b
Mouse recombinant GRO-b
Catalog # APOSBITC2013-1MG
Supplier:  Apollo Scientific
CAS Number:  
Mouse recombinant GRO-b
Catalog # APOSBITC2013-1MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pk:
    1 mg
  • Protein/peptide type:
    Recombinant
  • Species:
    Mouse
  • Protein synonyms:
    GRO-beta/MIP-2, CXCL2
  • Protein/peptide name:
    GRO-b

Specifications

About this item