Order Entry
Switzerland
ContactUsLinkComponent
Human recombinant exodus-2
Human recombinant exodus-2
Catalog # APOSBITC2005-1MG
Supplier:  Apollo Scientific
CAS Number:  
Human recombinant exodus-2
Catalog # APOSBITC2005-1MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pk:
    1 mg
  • Protein/peptide type:
    Recombinant
  • Species:
    Human
  • Protein synonyms:
    CCL21
  • Protein/peptide name:
    Exodus-2

Specifications

About this item