Orders LinkContactUsLinkComponent
b.safe Funnels 250
b.safe Funnels 250
Supplier:   Bohlender
b.safe Funnels 250
Supplier:  Bohlender
Certificates
b.safe Funnels 250
3 Options Available Below
Product Details & Documents
Funnel made of PE for liquid waste disposal. The funnel can be safely mounted on the waste container with its threaded connection that is safely fixed with the filling neck.
  • Hinged lid: With grip for easy opening and closing
  • Sieve: Big grip for safe grapping and removal of the sieve
  • Plug-in connection for filling tube: An additional filling tube can be plugged-in the funnel neck
A stainless steel sieve on the outlet restrains magnetic stirrer bars and other solid materials. It has a grip for easy removal for cleaning. The hinged lid reliably withholds vapours from the container.
Documents
Product Family Options
Product Information
  • Thread
    Availability
    Price
  • Funnel
    S60
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 218.00
    SFr. 218.00
    1 * 1 ST
       SFr. 218.00
  • Funnel
    G2"+Trisure2"
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 369.00
    SFr. 369.00
    1 * 1 ST
       SFr. 369.00
  • Funnel
    Mauser 2"
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 366.00
    SFr. 366.00
    1 * 1 ST
       SFr. 366.00
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Sieve for funnel 180
    Sieve for funnel 180
    590-0312
    Log in for availability
    1 * 1 ST - SFr. 35.80